Learn More
Vimentin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22031-1-AP-20UL
 View more versions of this product
Product Code. 16881575
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.
Specifications
Specifications
Vimentin
Polyclonal
Unconjugated
VIM
vimentin
Rabbit
Antigen Affinity Chromatography
RUO
22352, 7431, 81818
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08670, P20152, P31000
VIM
Vimentin Fusion Protein Ag16898
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vimentin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated