missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14878-1-AP-20UL
This item is not returnable.
View return policy
Description
The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.Specifications
VIPR1 | |
Polyclonal | |
Unconjugated | |
VIPR1 | |
HVR1, II, PACAP R 2, PACAP R2, PACAP type II receptor, RDC1, VAPC1, VIP R 1, VIPR, VIPR1, VIRG, VPAC1, VPCAP1R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7433 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P32241 | |
VIPR1 | |
VIPR1 Fusion Protein Ag6650 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VIPR1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction