Learn More
Viscosity Reference Standard Viscosity Standard Set, Cold Crank Simulator Type CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68. UKAS ISO 17025 / ISO 17034 Certified, 14 x 500 mL, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand:  Paragon Scientific VIS-CCS-14
Product Code. 16060500
  • 11750.00 SEK / Pack of 14
This item is not returnable. View return policy
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Specifications
14 x 500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Viscosity Reference Standard Viscosity Standard Set, Cold Crank Simulator Type CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68. UKAS ISO 17025 / ISO 17034 Certified, 14 x 500 mL, Paragon Scientific™ > Quantity: 14 x 500mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Nominal values quoted. Certified Values will vary from batch to batch.