Learn More
Viscosity Reference Standard Viscosity Standard Set, Cold Crank Simulator Type CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68. UKAS ISO 17025 / ISO 17034 Certified, 14 x 500 mL, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand: Paragon Scientific VIS-CCS-14
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
14 x 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.