Learn More
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets
Provide materials for maintenance of MLA D-Tipper™, Precision, and two-stroke pipetters
253.00 SEK
Specifications
For Use With | Pipettes |
---|---|
Material | Plastic |
Includes | Lubricant, seals and instructions |
Quantity | 1 Ea. |
Length (English) | 2 in. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
16405134
|
VistaLab™ Technologies
9031 |
5 to 30 ÎĽL |
253.00 SEK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
16415134
|
VistaLab™ Technologies
9032 |
31 to 120 ÎĽL |
253.00 SEK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
16425134
|
VistaLab™ Technologies
9033 |
121 to 200 ÎĽL |
253.00 SEK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
16435134
|
VistaLab™ Technologies
9034 |
201 to 350 ÎĽL |
253.00 SEK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
16445134
|
VistaLab™ Technologies
9035 |
351 to 1000 ÎĽL |
253.00 SEK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
Description
Each kit contains seals and instructions
Order Info
Lubricant sold separately (P/N 22-146-318)
Specifications
Pipettes | |
Lubricant, seals and instructions | |
2 in. | |
1 in. |
Plastic | |
1 Ea. | |
1 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets