Learn More
Heidolph instruments Viton™ Tubing
Use this tubing when you require excellent acid resistance at high temperatures. Heidolph™ Viton™ Tubing is smooth, biocompatible and inert.
795.00 SEK - 3255.00 SEK
Product Code | Brand | Tubing Size | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Thickness (Metric) | Price | Quantity & Availability | ||||
10504172
|
Heidolph instruments
525-52000-00 |
7.9 | 2.5 mm |
3255.00 SEK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
10065751
|
Heidolph instruments
525-53000-00 |
0.8 | 1.6 mm |
795.00 SEK
1m |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
10689722
|
Heidolph instruments
525-54000-00 |
1.7 | 1.6 mm |
855.00 SEK
1m |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
Description
- Low gas permeability.
- Resistant to solvents and corrosives at high temperatures.
Specifications
Autoclavable | |
Non-sterile |