Läs mer
Poulten & Graf GmbH Volac™ Polystop™ Clear Dropping Bottles
Produktkod. 12308208
Kapacitet (metrisk):
30 mL
50 mL
100 mL
Färg:
Amber
Clear
Förpackningsstorlek:
Paket med 10
Varje
Produktkod. 12308208
Brand: Poulten & Graf GmbH L188/A/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Manufactured from soda lime glass. Poulten Graf™ Volac™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable 3-piece plastic stopper.
  • Stopper consists of a strong ungraduated glass pipette, a durable vinyl teat and a PP-stopper
  • Available in clear or amber glass in a variety of sizes
  • Narrow neck construction
Specifikationer
Kapacitet (metrisk) 30 mL
Färg Clear
Material Glass
Stopparnummer 16/16
Produkttyp Dropping Bottle
Förslutningstyp Stopper