Learn More
VPS18 Mouse anti-Human, Unconjugated, Clone: 2E10G11, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | VPS18 |
---|---|
Clone | 2E10G11 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840306
|
Proteintech
67590-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16830306
|
Proteintech
67590-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The VPS18 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VPS18. This antibody recognizes human, rat antigen. The VPS18 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
VPS18 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
57617 | |
Primary |
2E10G11 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.