missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS37B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15653-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. May be involved in cell growth and differentiation.Specifications
VPS37B | |
Polyclonal | |
Unconjugated | |
VPS37B | |
ESCRT I complex subunit VPS37B, hVps37B, VPS37B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79720 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H9H4 | |
VPS37B | |
VPS37B Fusion Protein Ag8256 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |