Learn More
VPS45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12616-1-AP-150UL
 View more versions of this product
Product Code. 16880424
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators.
Specifications
Specifications
VPS45
Polyclonal
Unconjugated
VPS45
h VPS45, H1, H1VPS45, hlVps45, VPS45, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
Rabbit
Antigen Affinity Chromatography
RUO
11311, 22365, 64516
-20°C
Liquid
Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08700, P97390, Q9NRW7
Vps45
VPS45 Fusion Protein Ag3289
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Title
VPS45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated