Learn More
VSNL1 Mouse anti-Human, Unconjugated, Clone: 2C6C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67134-1-IG-20UL
 View more versions of this product
Product Code. 16889585
  • 1645.54 SEK / 20µL
Description
The VSNL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VSNL1. This antibody recognizes human, mouse, rat, pig antigen. The VSNL1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
VSNL1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
2C6C3
Unconjugated
20 μL
7447
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VSNL1 Mouse anti-Human, Unconjugated, Clone: 2C6C3, Proteintech > 20 μL