missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTI1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12354-1-AP-20UL
This item is not returnable.
View return policy
Description
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.Specifications
VTI1A | |
Polyclonal | |
Unconjugated | |
VTI1A | |
MVti1, Vti1 rp2, VTI1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
143187, 53611, 65277 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O89116, Q96AJ9, Q9JI51 | |
Vti1a | |
VTI1A Fusion Protein Ag3018 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VTI1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction