missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | VTN |
---|---|
Clone | 1D2C8 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898235
|
Proteintech
66398-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888235
|
Proteintech
66398-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.Specifications
VTN | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN | |
VTN | |
IgG1 | |
Protein G | |
Antibody |
1D2C8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Rabbit, Pig, Human | |
P04004, P22458, P48819 | |
100009128, 397192, 7448 | |
VTN Fusion Protein Ag8443 | |
Primary | |
-20°C | |
VTN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VTN Mouse anti-Human, Porcine, Rabbit, Clone: 1D2C8, Proteintech