Learn More
Macherey-Nagel™ Wash Buffer NT3 Concentrate
Concentrated ethanolic wash buffer for removing contaminations from bound DNA when using NucleoSpin™ and NucleoTraP™ PCR Clean-up kits. To prepare working buffer solution, add 25 mL concentrate to 100 mL ethanol (yields 125 mL working solution).
1540.00 SEK - 1860.00 SEK
Specifications
Color | Colorless |
---|---|
Concentration | 5X (50 mL concentrate yields 125 mL working solution) |
For Use With (Equipment) | NucleoTraP™ and NucleoSpin™ PCR clean-up kits |
pH | 7.0 to 8.0 |
Packaging | Concentrated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15880928
|
Macherey-Nagel™
740598A |
100 mL |
1540.00 SEK
100mL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899097
|
Macherey-Nagel™
740598.1 |
200 mL |
1860.00 SEK
200mL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NucleoTraP™ and NucleoSpin™ PCR clean-up kits allow DNA fragments to be purified from enzymatic reactions, such as PCR, as well as from agarose gels. Compatible kits include:
- NucleoSpin™ Gel and PCR Clean-up (standard, Midi, Maxi)
- NucleoSpin™ 8/96 PCR Clean-up (full and core kit)
- NucleoTraP™ CR and NucleoTraP™
Specifications
Colorless | |
NucleoTraP™ and NucleoSpin™ PCR clean-up kits | |
Concentrated | |
Room Temperature (18°C to 25°C) | |
Purification (clean up) of DNA |
5X (50 mL concentrate yields 125 mL working solution) | |
7.0 to 8.0 | |
Odorless | |
Liquid | |
Wash buffer |