Learn More
WBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11042-1-AP-20UL
 View more versions of this product
Product Code. 16887843
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The globular WW domain, named for the conserved tryptophan residues in the protein motif present in various structural and regulatory proteins, is known to play a role in the mediation of protein-protein interactions. This gene encodes a ligand of the WW domain of the Yes kinase-associated protein. Readthrough transcription of the neighboring upstream gene, which encodes INO80 complex subunit B, into this gene generates a non-coding transcript.
Specifications
Specifications
WBP1
Polyclonal
Unconjugated
WBP1
WBP 1, WBP1, WW domain binding protein 1
Rabbit
Antigen Affinity Chromatography
RUO
23559
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96G27
WBP1
WBP1 Fusion Protein Ag1478
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WBP1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated