missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBSCR16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | WBSCR16 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892184
|
Proteintech
13796-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882184
|
Proteintech
13796-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein containing regulator of chromosome condensation 1-like repeats. The encoded protein may function as a guanine nucleotide exchange factor. This gene is located in a region of chromosome 7 that is deleted in Williams-Beuren syndrome, a multisystem developmental disorder. Alternative splicing results in multiple transcript variants. Orthologs mouse allSpecifications
WBSCR16 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q96I51, Q9CYF5 | |
360796, 81554, 94254 | |
WBSCR16 Fusion Protein Ag4619 | |
Primary | |
-20°C | |
RCC1L |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
wbs16, WBSCR16 | |
Rcc1l | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WBSCR16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech