Learn More
WDR18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15165-1-AP-150UL
 View more versions of this product
Product Code. 16884224
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
WD repeat domain 18 (WDR18) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WD repeats are minimally conserved regions of approximately 40 amino acids that commonly form a tertiary propeller structure. WDR18 is a 432 amino acid protein that contains six WD repeats and is encoded by a gene located on human chromosome 19. WDR18 may play a role during development. The complex PELP1-TEX10-WDR18 is required for the maturation and nucleolar release of the large ribosomal subunit.
Specifications
Specifications
WDR18
Polyclonal
Unconjugated
WDR18
R32184_1, WD repeat domain 18, WDR18
Rabbit
Antigen Affinity Chromatography
RUO
57418
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BV38
WDR18
WDR18 Fusion Protein Ag7282
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WDR18 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated