missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20483-1-AP-150UL
This item is not returnable.
View return policy
Description
Proteintech's WDR32 antibody 20483-1-AP primarily detects WDR32 isoform 1 with a theoretical MW of 61 kDa.
DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.Specifications
WDR32 | |
Polyclonal | |
Unconjugated | |
DCAF10 | |
DCAF10, WD repeat domain 32, WDR32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
242418, 313242, 79269 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AKB9, Q5QP82 | |
Dcaf10 | |
WDR32 Fusion Protein Ag14360 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WDR32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction