Learn More
WDR33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22614-1-AP-150UL
 View more versions of this product
Product Code. 16882005
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody recognizes the full length WDR33 protein.
WDR33 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination.This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Specifications
WDR33
Polyclonal
Unconjugated
WDR33
NET14, WD repeat domain 33, WDC146, WDR33
Rabbit
Antigen Affinity Chromatography
RUO
55339
-20°C
Liquid
Immunoprecipitation, Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9C0J8
WDR33
WDR33 Fusion Protein Ag18382
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WDR33 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated