missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20916-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 55 kDa, and this antibody specifically recognizes the 55 kDa protein.
WDR37 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes.Specifications
WDR37 | |
Polyclonal | |
Unconjugated | |
WDR37 | |
KIAA0982, RP11 529L18.2, WD repeat domain 37, WDR37 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
207615, 22884, 307075 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CBE3, Q9Y2I8 | |
Wdr37 | |
WDR37 Fusion Protein Ag15112 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WDR37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction