missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR74 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20631-1-AP-150UL
This item is not returnable.
View return policy
Description
The yeast nucleolar protein NOP7 is necessary for the maturation of 66S preribosomes and interacts with numerous other proteins. One such protein is an essential, conserved WD repeat protein, NOP seven-associated protein 1 (NSA1), that is also required for the yeast 66S ribosome assembly. NSA1 is also associated with the AAA ATPase Rix7, and release of NSA1 from a novel late nucleolar pre-60S requires the Rix7 function. NSA1 has also been found upregulated in mammalian cancer cells, suggesting it may also play a role in cell proliferation.Specifications
WDR74 | |
Polyclonal | |
Unconjugated | |
WDR74 | |
NOP seven associated protein 1, NSA1, WD repeat domain 74, WDR74 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54663 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6RFH5 | |
WDR74 | |
WDR74 Fusion Protein Ag14617 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WDR74 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction