Learn More
Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Sheets and Rolls
100% pure nitrocellulose to ensure the highest binding capacity possible
3393.00 SEK
Specifications
Width (Metric) | 3 m |
---|---|
For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
Binding Capacity | High |
Quantity | 1 Each |
Format | Roll |
Product Code | Brand | Length (Metric) | Width (Metric) | Format | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Format | Quantity | Price | Quantity & Availability | |||||
11935104
|
Whatman™
10401197 |
20 cm | 3 m | Roll | 1 Each | N/A | ||||||
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
- Also available in circular formats
Specifications
3 m | |
High | |
Roll | |
0.45 ÎĽm |
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection | |
1 Each | |
Nitrocellulose |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Sheets and Rolls