Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Provide maximum surface between culture and glass flask
Brand: DWK Life Sciences 356915
1806.68 SEK valid until 2025-12-31
Use promo code "24329" to get your promotional price.
This item is not returnable.
View return policy
Description
Provide maximum surface between culture and glass flask
- PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
- Silicone rubber-lined cap
- Autoclavable
Specifications
Silicone Rubber | |
Flask only | |
PTFE Resin | |
38-430 |
50 mL | |
15-415 | |
Autoclavable |
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket > 50mL
Spot an opportunity for improvement?Share a Content Correction