Learn More
DWK Life Sciences Wheaton™ Connection Connectors
Safely joins two exterior threaded glass components without hooks, springs or clamps
Brand: DWK Life Sciences 125482
Additional Details : Weight : 0.45360kg
Description
- Screw cap configuration creates a grease-free, vacuum- tight seal
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Available with white polypropylene or black phenolic caps
- Phenolic connectors have a working temperature of up to 180
Specifications
Screw Thread Connector | |
Borosilicate Glass | |
Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere | |
Screw cap |
USP Type I, ASTM E438 Type I, Class A | |
Micro soxhlet extraction apparatus | |
12-425 and 24-400 | |
Polypropylene |