Learn More
DWK Life Sciences Wheaton™ Connection Connectors
Safely joins two exterior threaded glass components without hooks, springs or clamps
2775.00 SEK - 2900.00 SEK
Specifications
Product Type | Screw Thread Connector |
---|---|
Certifications/Compliance | USP Type I, ASTM E438 Type I, Class A |
Material | Borosilicate Glass |
For Use With | Micro soxhlet extraction apparatus |
For Use With (Application) | Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
12425442
|
DWK Life Sciences
125482 |
12-425 and 24-400 |
2775.00 SEK
Pack of 6 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
12445442
|
DWK Life Sciences
125488 |
20-400 and 24-400 |
2900.00 SEK
Pack of 6 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Description
- Screw cap configuration creates a grease-free, vacuum- tight seal
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Available with white polypropylene or black phenolic caps
- Phenolic connectors have a working temperature of up to 180
Specifications
Screw Thread Connector | |
Borosilicate Glass | |
Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere | |
Polypropylene |
USP Type I, ASTM E438 Type I, Class A | |
Micro soxhlet extraction apparatus | |
Screw cap |