Learn More
WNK4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22326-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16881855
  • 4721.67 SEK / 150µL
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB.
Specifications
Specifications
WNK4
Polyclonal
Unconjugated
WNK4
PHA2B, PRKWNK4, WNK4
Rabbit
Antigen Affinity Chromatography
RUO
65266, 69847
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80UE6, Q96J92
Wnk4
WNK4 Fusion Protein Ag17773
150 μL
Primary
Human, Mouse