Learn More
WNT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27935-1-AP-20UL
 View more versions of this product
Product Code. 16885905
  • 1645.54 SEK / 20µL
Description
The WNT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNT1. This antibody recognizes human, mouse antigen. The WNT1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
WNT1
Polyclonal
Unconjugated
20 μL
7471
IgG
Western Blot, Immunohistochemistry
300 μg/mL
Rabbit
Primary
Human, Mouse
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WNT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL