Learn More
WSB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12124-2-AP-150UL
 View more versions of this product
Product Code. 16889643
  • 4652.90 SEK / 150µL
Description
This gene encodes a member of the WD-protein subfamily. The encoded protein contains five WD-repeats spanning most of the protein and an SOCS box in the C-terminus. The SOCS box may act as a bridge between specific substrate-binding domains and E3 ubiquitin protein ligases.
Specifications
Specifications
WSB2
Polyclonal
Unconjugated
WSB2
CS box containing WD protein, SBA2, WSB 2, WSB2
Rabbit
Antigen Affinity Chromatography
RUO
288692, 55884, 59043
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O54929, Q9NYS7
Wsb2
WSB2 Fusion Protein Ag2768
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WSB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated