missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | WTAP |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886423
|
Proteintech
10200-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876423
|
Proteintech
10200-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wilms′ tumor (WT) is an embryonal maligcy of the kidney that affects 1 in 10,000 infants and is observed in both sporadic and inherited forms. The Wilms′ tumor protein (WT1) binds the DNA sequence GCGGGGGCG, a recognition element common to the early growth response (Egr) family of Zn2+ finger transcriptional activators, and functions as a transcriptional repressor. WTAP (Wilms tumor 1-associating protein) is a ubiquitously expressed nuclear protein that interacts with WT1 and may be involved in regulating mRNA splicing. WTAP is found in nuclear speckles, where it regulates the G2/M cell cycle transition by binding to the 3′ UTR of cyclin A2, thus enhancing its stability. Additionally, WTAP inhibits expression of WT1 target genes and is able to impair the ability of WT1 to bind DNA. Two isoforms of WTAP exist due to alternative splicing events.Specifications
WTAP | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q15007, Q9ER69 | |
499020, 60532, 9589 | |
WTAP Fusion Protein Ag0259 | |
Primary | |
-20°C | |
WTAP |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Female lethal(2)D homolog, hFL(2)D, KIAA0105, WT1 associated protein, WTAP | |
Wtap | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech