Learn More
WWP2 Mouse anti-Human, Unconjugated, Clone: 2C9B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67274-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The WWP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WWP2. This antibody recognizes human, rat antigen. The WWP2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
WWP2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Rat, Human |
ELISA, Immunohistochemistry, Western Blot | |
2C9B4 | |
Unconjugated | |
20 μL | |
11060 | |
IgG1 |