Learn More
WWP2 Mouse anti-Human, Unconjugated, Clone: 2C9B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67274-1-IG-150UL
 View more versions of this product
Product Code. 16879805
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The WWP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WWP2. This antibody recognizes human, rat antigen. The WWP2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
WWP2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunohistochemistry, ELISA
2C9B4
Unconjugated
150 μL
11060
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WWP2 Mouse anti-Human, Unconjugated, Clone: 2C9B4, Proteintech > 150 μL