Learn More
Invitrogen™ Human Leptin ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand: Invitrogen™ 900-K90K
This item is not returnable.
View return policy
Description
Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
Specifications
P41159 | |
Lot-specific | |
ELISA | |
3952 | |
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 45 mins |
Lot-specific | |
HRP | |
Colorimetric Microplate Reader | |
Lep | |
HRP | |
RUO | |
Leptin | |
24 hrs 30 mins |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human Leptin ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP
Spot an opportunity for improvement?Share a Content Correction