missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | XK |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850754
|
Proteintech
12830-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840754
|
Proteintech
12830-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This locus controls the synthesis of the Kell blood group ′precursor substance′ (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. XK has structural characteristics of prokaryotic and eukaryotic membrane transport proteins. This locus controls the synthesis of the Kell blood group ′precursor substance′ (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. The encoded protein has structural characteristics of prokaryotic and eukaryotic membrane transport proteins.Specifications
XK | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P51811, Q5GH61, Q9QXY7 | |
22439, 497078, 7504 | |
XK Fusion Protein Ag3550 | |
Primary | |
-20°C | |
XK |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Kell complex 37 kDa component, KX, Kx antigen, Membrane transport protein XK, X1k, XK, XK related protein 1, XKR1, XRG1 | |
Xk | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
XK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech