missing translation for 'onlineSavingsMsg'
Learn More

XPR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen XPR1
Concentration 0.15 mg/mL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16892694
View Documents
Proteintech
14174-1-AP-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16882694
View Documents
Proteintech
14174-1-AP-150UL
150 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
Specifications

Specifications

XPR1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
Q9UBH6, Q9Z0U0
19775, 289424, 9213
XPR1 Fusion Protein Ag5373
Primary
-20°C
XPR1
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Protein SYG1 homolog, SYG1, X receptor, X3, XPR1, XR
Xpr1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
XPR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.