Learn More
XRCC3 Mouse anti-Human, Unconjugated, Clone: 3A12F9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67222-1-IG-150UL
 View more versions of this product
Product Code. 16879745
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
The XRCC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human XRCC3. This antibody recognizes human, mouse antigen. The XRCC3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
XRCC3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry
3A12F9
Unconjugated
150 μL
7517
IgG2b
Product Title
XRCC3 Mouse anti-Human, Unconjugated, Clone: 3A12F9, Proteintech > 150 μL