Learn More
YPEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17743-1-AP-150UL
 View more versions of this product
Product Code. 16887694
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
YPEL1 (yippee-like 1) is thought to play a role in the epithelioid conversion of fibroblasts and the regulation of the cellular morphology. It is a 119 amino acid protein that localizes to the nucleus and belongs to a family of yippee-like proteins that include YPEL2 - 5, all of which localize to the centrosome and nucleus and are widely expressed in both adult and fetal tissue. The family of human YPEL proteins share a high degree of sequence homology with their rodent homologs, suggesting a conserved function between species. YPEL1 might be an important factor during the development and maligt transformation of tissues.
Specifications
Specifications
YPEL1
Polyclonal
Unconjugated
YPEL1
FKSG3, Protein yippee like 1, yippee like 1 (Drosophila), YPEL1
Rabbit
Antigen Affinity Chromatography
RUO
100360867, 106369, 29799
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60688, Q9ESC7
Ypel1
YPEL1 Fusion Protein Ag11993
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
YPEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated