Learn More
YTHDF2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26771-1-AP-150UL
 View more versions of this product
Product Code. 16875135
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The YTHDF2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human YTHDF2. This antibody recognizes human, mouse antigen. The YTHDF2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
YTHDF2
Polyclonal
Unconjugated
150 μL
51441
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
200 μg/mL
Rabbit
Primary
Human, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
YTHDF2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 150 μL