Learn More
YTHDF2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26771-1-AP-150UL
Description
The YTHDF2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human YTHDF2. This antibody recognizes human, mouse antigen. The YTHDF2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
YTHDF2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
51441 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |