Learn More
YWHAB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10936-1-AP-20UL
 View more versions of this product
Product Code. 16887653
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This antibody was raised against the N-terminal region of human YWHAB.
14-3-3, a family of acidic and soluble proteins, highly conserved in amino acid sequences from yeast to mammals, is expressed in alleukaryotic cells. Seven isoforms encoded by seven distinct genes are identified in mammals and forms homo-and hetero-dimericcup-shaped structures. As 14-3-3 is interacted with more than 100 binding partners, it regulates key proteins involved in various biological processes such as signal trans-duction, cell cycle, transcriptional control, cell proliferation, apoptosis, and ion channel physiology. Most 14-3-3 requires phosphorylation of serine or threonine residues in the target sequence. This protein is abundantly expressed in the brain and has been detected in the cerebrospinal fluid of patients with different neurological disorders.
Specifications
Specifications
YWHAB
Polyclonal
Unconjugated
YWHAB
14 3 3, 14 3 3 alpha, 14 3 3 protein beta/alpha, YWHAB
Rabbit
Antigen Affinity Chromatography
RUO
54401, 56011, 7529
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P31946, P35213, Q9CQV8
Ywhab
YWHAB Fusion Protein Ag1383
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
YWHAB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated