Läs mer
(Z)- Disperse Yellow 39, TRC
Produktkod. 30179446
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30179446 2.5 g 2.5g
30179577 1 g -
30179664 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179446 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4960352.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38072-53-6
Molekylformel C17H16N2O
Molekylvikt (g/mol) 264.32
Synonym (2Z)-2-[[4-(Dimethylamino)phenyl]methylene]-1,2-dihydro-3H-indol-3-one
LEDER CN(C)c1ccc(C=C2Nc3ccccc3C2=O)cc1
Specifikationer
Kemiskt namn eller material (Z)- Disperse Yellow 39
Kvantitet 2.5 g
InChI formel InChI=1S/C17H16N2O/c1-19(2)13-9-7-12(8-10-13)11-16-17(20)14-5-3-4-6-15(14)18-16/h3-11,18H,1-2H3/b16-11+