missing translation for 'onlineSavingsMsg'
Learn More

ZADH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14679-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883434

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZADH2 (zinc binding alcohol dehydrogenase domain containing 2) is a 377 amino acid protein that belongs to the zinc-containing alcohol dehydrogenase family and is encoded by a gene which maps to human chromosome 18. Chromosome 18 houses over 300 protein-coding genes and contains nearly 76 million bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.
Specifications

Specifications

ZADH2
Polyclonal
Unconjugated
ZADH2
ZADH2
Rabbit
Antigen Affinity Chromatography
RUO
225791, 284273, 291403
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGC4, Q8N4Q0
ZADH2
ZADH2 Fusion Protein Ag6390
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers