missing translation for 'onlineSavingsMsg'
Learn More

ZAP70 Mouse anti-Human, Mouse, Porcine, Clone: 1A11G9, Proteintech

Mouse Monoclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen ZAP70
Clone 1A11G9
Concentration 1.37 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887195
View Documents
Proteintech
60200-1-IG-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16877195
View Documents
Proteintech
60200-1-IG-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interacts with phosphorylated C-terminal heptapeptide repeat domain (CTD) of the largest RNA polymerase II subunit POLR2A, and participates in dephosphorylation of the CTD. Transcriptional regulator which enhances expression of CCND1. Promotes binding of RNA polymerase II to the CCDN1 promoter and to the termination region before the poly-A site but decreases its binding after the poly-A site. Prevents RNA polymerase II from reading through the 3′ end termination site and may allow it to be recruited back to the promoter through promotion of the formation of a chromatin loop. Also enhances the transcription of a number of other cell cycle-related genes including CDK2, CDK4, CDK6 and cyclin-E but not CDKN1A, CDKN1B or cyclin-A. Promotes cell proliferation.
Specifications

Specifications

ZAP70
1.37 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
70 kDa zeta associated protein, SRK, STD, Syk related tyrosine kinase, Tyrosine protein kinase ZAP 70, TZK, ZAP 70, ZAP70
ZAP70
IgG2a
Protein A
Antibody
1A11G9
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Pig, Mouse
P43403, P43404
100739325, 22637, 7535
ZAP70 Fusion Protein Ag7943
Primary
-20°C
ZAP70
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ZAP70 Mouse anti-Human, Mouse, Porcine, Clone: 1A11G9, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.