missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCRB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25629-1-AP-20UL
This item is not returnable.
View return policy
Description
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.Specifications
ZCRB1 | |
Polyclonal | |
Unconjugated | |
ZCRB1 | |
MADP 1, MADP1, RBM36, U11/U12 31K, U11/U12 snRNP 31 kDa protein, ZCCHC19, ZCRB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67197, 85437 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TBF4, Q9CZ96 | |
ZCRB1 | |
ZCRB1 Fusion Protein Ag22095 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZCRB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction