missing translation for 'onlineSavingsMsg'
Learn More

ZFP64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17187-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887034

  • 1355.00 SEK / 20µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications

Specifications

ZFP64
Polyclonal
Unconjugated
ZFP64
Zfp 64, ZFP64, Zinc finger protein 338, ZNF338
Rabbit
Antigen Affinity Chromatography
RUO
22722, 311661, 55734
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99KE8, Q9NPA5
Zfp64
ZFP64 Fusion Protein Ag10971
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers