missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate Hexahydrate, purum p.a., ≥99% (KT), Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.

Specifications
Specifications
Melting Point | 38.9°C |
Color | Colorless |
Quantity | 1 kg |
Linear Formula | Zn(NO3)2 · 6H2O |
UN Number | UN1514 |
IUPAC Name | zinc;dinitrate;hexahydrate |
Formula Weight | 297.49g/mol |
Percent Purity | ≥99% (KT) |
Odor | Weak |
Packaging | Poly Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc Nitrate Hexahydrate, purum p.a., ≥99% (KT), Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction