Learn More
RETSCH Zirconium Oxide Grinding Sets
Designed for extremely quick, loss-free and reproducible grinding of medium-hard, brittle and fibrous materials to analytical fineness. Retsch™ Zirconium Oxide Grinding Sets are ideal for use in the building materials sector (cement), in geology, mineralogy, metallurgy and in power plants.
Brand:  RETSCH 01.462.0192
 View more versions of this product
Product Code. 10740093
  • 93525.00 SEK / Each
Only null left
Estimated Shipment: 19-08-2025
Description
  • For use with RS 100 and RS 200 Vibratory Disc Mills
Specifications
Specifications
Grinding Set
100 mL
For RS200
Zirconium Oxide
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Zirconium Oxide Grinding Sets > Capacity: 100mL