Learn More
ZMAT2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25819-1-AP-20UL
 View more versions of this product
Product Code. 16884375
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Specifications
ZMAT2
Polyclonal
Unconjugated
ZMAT2
zinc finger, matrin type 2, ZMAT2
Rabbit
Antigen Affinity Chromatography
RUO
153527, 679898
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96NC0
Zmat2
ZMAT2 Fusion Protein Ag22909
20 μL
Primary
Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZMAT2 Rabbit anti-Human, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated