missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF211 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25458-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May be involved in transcriptional regulation.Specifications
ZNF211 | |
Polyclonal | |
Unconjugated | |
ZNF211 | |
C2H2 25, CH2H2 25, zinc finger protein 211, Zinc finger protein C2H2 25, ZNF 25, ZNF211, ZNFC25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10520 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13398 | |
ZNF211 | |
ZNF211 Fusion Protein Ag22135 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |