missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF211 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ZNF211 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884035
|
Proteintech
25458-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874035
|
Proteintech
25458-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
May be involved in transcriptional regulation.Specifications
ZNF211 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q13398 | |
10520 | |
ZNF211 Fusion Protein Ag22135 | |
Primary | |
-20°C | |
ZNF211 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C2H2 25, CH2H2 25, zinc finger protein 211, Zinc finger protein C2H2 25, ZNF 25, ZNF211, ZNFC25 | |
ZNF211 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |