Learn More
ZNF215 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25502-1-AP-20UL
 View more versions of this product
Product Code. 16884065
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZNF215
Polyclonal
Unconjugated
ZNF215
BAZ 2, BAZ2, zinc finger protein 215, ZKSCAN11, ZNF215
Rabbit
Antigen Affinity Chromatography
RUO
7762
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UL58
ZNF215
ZNF215 Fusion Protein Ag22026
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF215 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated