missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF238 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ZNF238 |
---|---|
Concentration | 0.54 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890584
|
Proteintech
12714-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880584
|
Proteintech
12714-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF238 is the sequence-specific DNA-binding protein with transcriptional repression activity. ZNF238 binds the consensus DNA sequence 5′-[AC]ACATCTG[GT][AC]-3′ which contains the E box core. ZNF238 may play a role in the organization of chromosomes in the nucleus.C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.Specifications
ZNF238 | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q99592, Q9JKY3, Q9WUK6 | |
10472, 30928, 64619 | |
ZNF238 Fusion Protein Ag3406 | |
Primary | |
-20°C | |
ZBTB18 |
0.54 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
58 kDa repressor protein, C2H2 171, RP58, TAZ 1, TAZ1, Transcriptional repressor RP58, ZBTB18, ZFP238, zinc finger protein 238, Zinc finger protein C2H2 171, ZNF238 | |
Zbtb18 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF238 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech