missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF266 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | ZNF266 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878403
|
Proteintech
11374-1-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 12-06-2023 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16888403
|
Proteintech
11374-1-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 12-06-2023 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.Specifications
ZNF266 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q14584 | |
10781 | |
ZNF266 Fusion Protein Ag1933 | |
Primary | |
-20°C | |
ZNF266 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HZF1, KIAA2007, zinc finger protein 266, Zinc finger protein HZF1, ZNF266 | |
ZNF266 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |